Assay Details
| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138660.2 | NP_619601.2 | 6 - 7 | 833 | 91 | |
| GenBank mRNA | AF526276.1 | - | 6 - 7 | 807 | 91 | |
| AJ292072.1 | - | 1047 | 91 | |||
| AK139608.1 | - | 6 - 7 | 872 | 91 | ||
| BC034533.1 | - | 3 - 4 | 477 | 91 | ||
| BC060672.1 | - | 6 - 7 | 661 | 91 | ||
| BC141296.1 | - | 6 - 7 | 946 | 9055908 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casc3
Genomic Map