Assay Details
| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138660.2 | NP_619601.2 | 7 - 8 | 1511 | 64 | |
| GenBank mRNA | AF526276.1 | - | 7 - 8 | 1485 | 64 | |
| AJ292072.1 | - | 1725 | 64 | |||
| AK037399.1 | - | 1 - 2 | 105 | 64 | ||
| AK139608.1 | - | 7 - 8 | 1550 | 64 | ||
| AK145886.1 | - | 7 - 8 | 1285 | 64 | ||
| AK183690.1 | - | 1 - 2 | 370 | 64 | ||
| BC060672.1 | - | 7 - 8 | 1339 | 64 | ||
| BC141296.1 | - | 7 - 8 | 1624 | 9055908 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casc3
Genomic Map