Assay Details
| Gene Symbol: Igfbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010517.3 | NP_034647.1 | 2 - 3 | 733 | 102 | |
| GenBank mRNA | AK031212.1 | - | 2 - 3 | 750 | 102 | |
| AK150229.1 | - | 2 - 3 | 746 | 102 | ||
| AK165688.1 | - | 751 | 102 | |||
| BC019836.1 | - | 2 - 3 | 733 | 5123738 | 102 | |
| X81582.1 | - | 2 - 3 | 682 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp4
Genomic Map