Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177390.3 | NP_796364.2 | 10 - 11 | 1524 | 86 | |
| GenBank mRNA | AK049374.1 | - | 10 - 11 | 1525 | 86 | |
| BC039700.1 | - | 5 - 6 | 656 | 86 | ||
| BC150765.1 | - | 10 - 11 | 1390 | 9087676 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1d
Genomic Map