Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177390.3 | NP_796364.2 | 14 - 15 | 1977 | 87 | |
| GenBank mRNA | AK049374.1 | - | 14 - 15 | 1978 | 87 | |
| AK148112.1 | - | 1 - 2 | 96 | 87 | ||
| BC150765.1 | - | 14 - 15 | 1843 | 9087676 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1d
Genomic Map