Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177390.3 | NP_796364.2 | 20 - 21 | 2936 | 76 | |
| GenBank mRNA | AK148112.1 | - | 7 - 8 | 1055 | 76 | |
| BC034071.1 | - | 6 - 7 | 916 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo1d
Genomic Map