Assay Details
| Gene Symbol: Tmem98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029537.1 | NP_083813.1 | 3 - 4 | 362 | 118 | |
| GenBank mRNA | AK018332.1 | - | 2 - 3 | 157 | 118 | |
| AK148132.1 | - | 3 - 4 | 361 | 118 | ||
| AK153960.1 | - | 3 - 4 | 352 | 118 | ||
| BC011208.1 | - | 3 - 4 | 362 | 4163101 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem98
Genomic Map