Assay Details
| Gene Symbol: Tmem98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029537.1 | NP_083813.1 | 5 - 6 | 517 | 61 | |
| GenBank mRNA | AK018332.1 | - | 4 - 5 | 312 | 61 | |
| AK148132.1 | - | 5 - 6 | 516 | 61 | ||
| AK153960.1 | - | 5 - 6 | 507 | 61 | ||
| BC011208.1 | - | 5 - 6 | 517 | 4163101 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem98
Genomic Map