Assay Details
| Gene Symbol: Serpinb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159748.1 | NP_001153220.1 | 5 - 6 | 865 | 82 | |
| NM_011459.4 | NP_035589.1 | 5 - 6 | 865 | 82 | ||
| GenBank mRNA | AK032659.1 | - | 5 - 6 | 864 | 82 | |
| AK036811.1 | - | 5 - 6 | 801 | 82 | ||
| AK048230.1 | - | 5 - 6 | 825 | 82 | ||
| AK148413.1 | - | 5 - 6 | 863 | 82 | ||
| BC025121.1 | - | 5 - 6 | 855 | 4487039 | 82 | |
| U96703.2 | - | 5 - 6 | 856 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027012 | AK036811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027012 | NM_011459.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027012 | AK032659.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42126 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140294 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178411 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027012 | AK036811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027012 | NM_011459.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027012 | AK032659.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20513541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140294 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04218280 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04218305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178411 | - | - |
| Whole Mouse Genome G4122A | A_52_P544885 | AK036811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544885 | NM_011459.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P544885 | AK032659.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521054 | U96703.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521054 | AK148413.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521054 | BC011076.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521054 | BC025121.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521054 | AK036811.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116190 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178411 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00995899 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00995900 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00208770 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00995897 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00208772 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00208768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00208787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20513592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00208770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00075493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00409814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00075518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00107744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00870757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00715020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00208783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00075510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00498972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00494893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00208793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00075503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00838443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00026210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00208776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20513562 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10349180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349176 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422776_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458882_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10349174 | - | - |
| Mouse Genome 430 2.0 Array | 1422776_at | - | - |
| Mouse Genome 430 2.0 Array | 1458882_at | - | - |
| Mouse Genome 430 2.0 Array | 1458882_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422776_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20725_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20725_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100008575.mm.1 | NM_001159748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008575.mm.1 | NM_001159748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008575.mm.1 | TC0100001031.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100008573.mm.1 | NM_001159748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008573.mm.1 | NM_001159748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008573.mm.1 | NM_011459.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008573.mm.1 | NM_011459.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100008573.mm.1 | TC0100001031.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422776_at | - | - |
| Mouse Expression Set 430 | 1422776_PM_at | - | - |
| Mouse Expression Set 430 | 1458882_at | - | - |
| Mouse Expression Set 430 | 1458882_PM_at | - | - |
| Murine Genome U74v2 Set | 167918_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167918_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422776_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422776_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422776_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422776_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458882_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458882_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 347755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4050465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050465 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223485 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223485 | - | - |
| mouse-6 v1.1 (Illumina) | scl17574.7_640-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050465 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17574.7_640-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050465 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2829250 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2829250 | - | - |

Serpinb8
Genomic Map