Assay Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170671.2 | NP_733771.2 | 17 - 18 | 2584 | 119 | |
| GenBank mRNA | AK029929.1 | - | 16 - 17 | 2760 | 119 | |
| AK166780.1 | - | 10 - 11 | 1732 | 119 | ||
| BC027327.1 | - | 13 - 14 | 1945 | 3991131 | 119 | |
| BC050808.1 | - | 6 - 7 | 1107 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mycbpap
Genomic Map