Assay Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170671.2 | NP_733771.2 | 4 - 5 | 493 | 69 | |
| GenBank mRNA | AK029929.1 | - | 4 - 5 | 422 | 69 | |
| BU840494.1 | - | 4 - 5 | 493 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029929.1 transcripts.
Set Membership: |
|

Mycbpap
Genomic Map