Assay Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026313.1 | NP_080589.1 | 3 - 4 | 323 | 133 | |
| GenBank mRNA | AK014362.1 | - | 3 - 4 | 316 | 133 | |
| AK019464.1 | - | 3 - 4 | 376 | 133 | ||
| AK078090.1 | - | 3 - 4 | 370 | 133 | ||
| AK151228.1 | - | 3 - 4 | 371 | 133 | ||
| AK151839.1 | - | 3 - 4 | 371 | 133 | ||
| BC009092.1 | - | 3 - 4 | 323 | 3157532 | 133 | |
| EF410121.1 | - | 355 | 133 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Luc7l3
Genomic Map