Assay Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026313.1 | NP_080589.1 | 4 - 5 | 473 | 122 | |
| GenBank mRNA | AK014362.1 | - | 4 - 5 | 466 | 122 | |
| AK019464.1 | - | 4 - 5 | 526 | 122 | ||
| AK078090.1 | - | 4 - 5 | 520 | 122 | ||
| AK151228.1 | - | 4 - 5 | 521 | 122 | ||
| AK151839.1 | - | 4 - 5 | 521 | 122 | ||
| BC009092.1 | - | 4 - 5 | 473 | 3157532 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Luc7l3
Genomic Map