Assay Details
Gene Symbol: Rsad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013381.2 | NP_001013399.1 | 3 - 4 | 566 | 55 | |
GenBank mRNA | AK046706.1 | - | 3 - 4 | 571 | 55 | |
AK148401.1 | - | 3 - 4 | 561 | 55 | ||
BB645699.1 | - | 3 - 4 | 571 | 55 | ||
BC139274.1 | - | 3 - 4 | 509 | 8862296 | 55 | |
BC139276.1 | - | 3 - 4 | 509 | 8862298 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
oxidation-reduction process
catalytic activity
coproporphyrinogen oxidase activity
oxidoreductase activity
short-branched-chain-acyl-CoA dehydrogenase activity
very-long-chain-acyl-CoA dehydrogenase activity
4-methyloctanoyl-CoA dehydrogenase activity
naphthyl-2-methyl-succinyl-CoA dehydrogenase activity
2-methylhexanoyl-CoA dehydrogenase activity
propionyl-CoA dehydrogenase activity
thiol-driven fumarate reductase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
coenzyme F420-dependent 2,4,6-trinitrophenol reductase activity
coenzyme F420-dependent 2,4,6-trinitrophenol hydride reductase activity
coenzyme F420-dependent 2,4-dinitrophenol reductase activity
medium-chain-acyl-CoA dehydrogenase activity
porphyrin-containing compound biosynthetic process
oxidation-reduction process
catalytic activity
coproporphyrinogen oxidase activity
oxidoreductase activity
short-branched-chain-acyl-CoA dehydrogenase activity
very-long-chain-acyl-CoA dehydrogenase activity
4-methyloctanoyl-CoA dehydrogenase activity
naphthyl-2-methyl-succinyl-CoA dehydrogenase activity
2-methylhexanoyl-CoA dehydrogenase activity
propionyl-CoA dehydrogenase activity
thiol-driven fumarate reductase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
coenzyme F420-dependent 2,4,6-trinitrophenol reductase activity
coenzyme F420-dependent 2,4,6-trinitrophenol hydride reductase activity
coenzyme F420-dependent 2,4-dinitrophenol reductase activity
medium-chain-acyl-CoA dehydrogenase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033657 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033656 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033654 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033655 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | BC056485.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | AK046706.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | AK148401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | BC056485.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | AK046706.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | AK148401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12737 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513570 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401758 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4119 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660353 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101179 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109335 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | BC056485.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | AK046706.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | AK148401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | BC056485.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | AK046706.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | AK148401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033668 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033660 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28033644 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109335 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02399429 | - | - |
Whole Mouse Genome G4122A | A_51_P514913 | BC056485.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | AK046706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | AK148401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | BC056485.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | AK046706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | AK148401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P514913 | NM_001013381.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109335 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575212 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575216 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575220 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575215 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09313406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033654 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033655 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033657 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28033656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28033602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09281879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09575212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09218554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09383920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09319912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09570420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09521168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09218552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09521170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09695575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09738095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09424896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09344057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09417733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09653177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28033659 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390024 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390030 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390026 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390025 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390023 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390028 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390029 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390031 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420321_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437449_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440875_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390022 | - | - |
Mouse Genome 430 2.0 Array | 1420321_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437449_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437449_at | - | - |
Mouse Genome 430 2.0 Array | 1440875_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1420321_at | - | - |
Mouse Genome 430 2.0 Array | 1440875_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237926_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237926_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100032638.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032638.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032638.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032646.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032646.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032646.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032641.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032641.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032641.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032640.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032640.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032640.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032645.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032645.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032645.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032651.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032651.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032651.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032639.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032639.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032639.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032635.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032635.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032635.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032650.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032650.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032650.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032636.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032636.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032636.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032637.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032637.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032637.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032643.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032643.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032643.mm.1 | TC1100003566.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100032642.mm.1 | NM_001013381.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032642.mm.1 | NM_001013381.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100032642.mm.1 | TC1100003566.mm.1 | MTA TC |
Mouse Expression Set 430 | 1437449_at | - | - |
Mouse Expression Set 430 | 1440875_a_at | - | - |
Mouse Expression Set 430 | 1420321_at | - | - |
Mouse Expression Set 430 | 1420321_PM_at | - | - |
Mouse Expression Set 430 | 1437449_PM_at | - | - |
Mouse Expression Set 430 | 1440875_PM_a_at | - | - |
Murine Genome U74v2 Set | 140194_at | - | - |
Murine Genome U74v2 Set | 140194_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440875_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1420321_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1420321_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437449_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437449_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440875_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2592861 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2802168 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2802168 | - | - |
