Assay Details
| Gene Symbol: Rsad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013381.2 | NP_001013399.1 | 5 - 6 | 994 | 78 | |
| GenBank mRNA | AK046706.1 | - | 5 - 6 | 999 | 78 | |
| AK148401.1 | - | 5 - 6 | 990 | 78 | ||
| BC056485.1 | - | 2 - 3 | 324 | 78 | ||
| BC139274.1 | - | 5 - 6 | 937 | 8862296 | 78 | |
| BC139276.1 | - | 5 - 6 | 937 | 8862298 | 78 | |
| BC145594.1 | - | 5 - 6 | 918 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsad1
Genomic Map