Assay Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145828.3 | NP_665827.2 | 7 - 8 | 1571 | 69 | |
| GenBank mRNA | AK083482.1 | - | 7 - 8 | 1540 | 69 | |
| AK086804.1 | - | 7 - 8 | 1503 | 69 | ||
| AK150521.1 | - | 7 - 8 | 1524 | 69 | ||
| AK153332.1 | - | 7 - 8 | 1524 | 69 | ||
| AK158056.1 | - | 7 - 8 | 1571 | 69 | ||
| BC034082.1 | - | 6 - 7 | 1090 | 4239646 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylt2
Genomic Map