Assay Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053161.2 | NP_444391.1 | 3 - 4 | 249 | 71 | |
| GenBank mRNA | AB049648.1 | - | 3 - 4 | 244 | 71 | |
| AK143575.1 | - | 3 - 4 | 277 | 71 | ||
| BC030074.1 | - | 3 - 4 | 249 | 1514322 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl27
Genomic Map