Assay Details
| Gene Symbol: Nckap5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001884.1 | NP_001001884.1 | 9 - 10 | 3449 | 56 | |
| GenBank mRNA | AK048721.1 | - | 2 - 3 | 979 | 56 | |
| BC026468.1 | - | 2 - 3 | 763 | 56 | ||
| BC072572.1 | - | 9 - 10 | 3449 | 5719876 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5l
Genomic Map