Assay Details
| Gene Symbol: Nckap5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001884.1 | NP_001001884.1 | 11 - 12 | 3832 | 52 | |
| GenBank mRNA | AK048721.1 | - | 4 - 5 | 1362 | 52 | |
| BC026468.1 | - | 4 - 5 | 1146 | 52 | ||
| BC072572.1 | - | 11 - 12 | 3832 | 5719876 | 52 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5l
Genomic Map