Assay Details
| Gene Symbol: Fmnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011711.1 | NP_035841.1 | 12 - 13 | 1387 | 76 | |
| GenBank mRNA | AK129473.1 | - | 12 - 13 | 1306 | 76 | |
| AK139711.1 | - | 12 - 13 | 1370 | 76 | ||
| AK143308.1 | - | 12 - 13 | 1286 | 76 | ||
| AK155331.1 | - | 12 - 13 | 1404 | 76 | ||
| AK163361.1 | - | 12 - 13 | 1373 | 76 | ||
| AK170744.1 | - | 11 - 12 | 1217 | 76 | ||
| BC023797.1 | - | 11 - 12 | 1107 | 76 | ||
| BC085094.1 | - | 12 - 13 | 1284 | 76 | ||
| BC131961.1 | - | 12 - 13 | 1263 | 40130238 | 76 | |
| U92453.1 | - | 1 - 2 | 62 | 76 | ||
More Information
Set Membership: |
|

Fmnl3
Genomic Map