Assay Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007405.2 | NP_031431.2 | 13 - 14 | 2318 | 93 | |
| GenBank mRNA | AK147516.1 | - | 13 - 14 | 2318 | 93 | |
| BC145101.1 | - | 12 - 13 | 2151 | 93 | ||
| M93422.1 | - | 12 - 13 | 2250 | 93 | ||
More Information
Set Membership: |
|

Adcy6
Genomic Map