Assay Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044741.2 | NP_001038206.1 | 9 - 10 | 893 | 82 | |
| NM_001286226.1 | NP_001273155.1 | 8 - 9 | 1066 | 82 | ||
| NM_007581.3 | NP_031607.2 | 9 - 10 | 1147 | 82 | ||
| GenBank mRNA | AF498245.1 | - | 9 - 10 | 745 | 82 | |
| AF498246.1 | - | 9 - 10 | 746 | 82 | ||
| AK148088.1 | - | 8 - 9 | 1067 | 82 | ||
| BG085743.2 | - | 1 - 2 | 97 | 82 | ||
| BQ042307.1 | - | 7 - 8 | 566 | 82 | ||
| U20372.1 | - | 9 - 10 | 801 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacnb3
Genomic Map