Assay Details
| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081434.1 | NP_001074903.1 | 16 - 17 | 1891 | 86 | |
| GenBank mRNA | AK154677.1 | - | 16 - 17 | 1929 | 86 | |
| AK171585.1 | - | 16 - 17 | 1920 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047981 | AK171585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047981 | BC085242.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047981 | AK038088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047981 | NM_001081434.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579436 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15829 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104723 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047981 | AK171585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047981 | BC085242.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047981 | AK038088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047981 | NM_001081434.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07454618 | - | - |
| Whole Mouse Genome G4122A | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275354 | AK154677.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275354 | NM_001081434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511259 | AK017095.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09167714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09124147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09359946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09559750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09105026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09536032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09593726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09609897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09167705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09498530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09593730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09498522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09167708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09205303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09559754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09167716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09735704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09330246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09205307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09631285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09167718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380748 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380750 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380743 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428893_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380739 | - | - |
| Mouse Genome 430 2.0 Array | 1428893_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459966_at | - | - |
| Mouse Genome 430 2.0 Array | 1428893_at | - | - |
| Mouse Genome 430 2.0 Array | 1459966_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71240_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71240_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014109.mm.1 | NM_001081434.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014109.mm.1 | NM_001081434.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014109.mm.1 | TC1100001530.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014110.mm.1 | NM_001081434.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014110.mm.1 | NM_001081434.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014110.mm.1 | TC1100001530.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014111.mm.1 | NM_001081434.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014111.mm.1 | NM_001081434.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014111.mm.1 | TC1100001530.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428893_at | - | - |
| Mouse Expression Set 430 | 1428893_PM_at | - | - |
| Mouse Expression Set 430 | 1459966_PM_at | - | - |
| Mouse Expression Set 430 | 1459966_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428893_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459966_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428893_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459966_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 764673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 105900750 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2476272 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227728 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001341.1_93-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105900750 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105900750 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2476272 | - | - |

Osbpl7
Genomic Map