Assay Details
| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081434.1 | NP_001074903.1 | 19 - 20 | 2180 | 67 | |
| GenBank mRNA | AK017095.1 | - | 1 - 2 | 201 | 67 | |
| AK154677.1 | - | 19 - 20 | 2218 | 67 | ||
| AK171585.1 | - | 19 - 20 | 2209 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl7
Genomic Map