Assay Details
| Gene Symbol: Osbpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081434.1 | NP_001074903.1 | 6 - 7 | 633 | 64 | |
| GenBank mRNA | AK038088.1 | - | 1 - 2 | 1220 | 64 | |
| AK154677.1 | - | 6 - 7 | 667 | 64 | ||
| AK171585.1 | - | 6 - 7 | 659 | 64 | ||
| BC085242.1 | - | 6 - 7 | 868 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl7
Genomic Map