Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100012179.mm.1 |
NM_028589.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012179.mm.1 |
NM_028589.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012179.mm.1 |
TC1100001288.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012178.mm.1 |
NM_028589.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012178.mm.1 |
NM_028589.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012178.mm.1 |
TC1100001288.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100012176.mm.1 |
NM_028589.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012176.mm.1 |
NM_028589.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100012176.mm.1 |
TC1100001288.mm.1 |
MTA TC |