Assay Details
| Gene Symbol: 1700125H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028589.1 | NP_082865.1 | 5 - 6 | 702 | 108 | |
| GenBank mRNA | AK007281.1 | - | 4 - 5 | 424 | 108 | |
| BC048674.1 | - | 5 - 6 | 702 | 6774125 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032265 | NM_028589.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032265 | BC048674.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144115 | NM_028589.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144115 | AK007281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144115 | BC048674.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163771 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112838 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115135 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144115 | NM_028589.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144115 | AK007281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144115 | BC048674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032265 | NM_028589.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032265 | BC048674.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112838 | - | - |
| Whole Mouse Genome G4122A | A_51_P299858 | NM_028589.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299858 | AK007281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299858 | BC048674.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112838 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09164231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09070407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09364431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08982874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09658766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09097584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09664690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09161868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09191916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27981537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379882 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429873_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379880 | - | - |
| Mouse Genome 430 2.0 Array | 1429873_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429873_at | - | - |
| Mouse Genome 430 2.0 Array | 1447220_at | - | - |
| Mouse Genome 430 2.0 Array | 1447220_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73634_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73634_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100012182.mm.1 | NM_028589.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012182.mm.1 | NM_028589.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012182.mm.1 | TC1100001288.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012183.mm.1 | NM_028589.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012183.mm.1 | NM_028589.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012183.mm.1 | TC1100001288.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429873_PM_at | - | - |
| Mouse Expression Set 430 | 1429873_at | - | - |
| Murine Genome U74v2 Set | 171269_i_at | - | - |
| Murine Genome U74v2 Set | 171269_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168783_f_at | - | - |
| Murine Genome U74v2 Set | 168783_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429873_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429873_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 720490 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214957 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214957 | - | - |

1700125H20Rik
Genomic Map