Assay Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102565.1 | NP_001096035.1 | 3 - 4 | 461 | 78 | |
| GenBank mRNA | AK042102.1 | - | 3 - 4 | 462 | 78 | |
| AK143105.1 | - | 5 - 6 | 670 | 78 | ||
| AK144828.1 | - | 3 - 4 | 462 | 78 | ||
| BC050856.1 | - | 4 - 5 | 586 | 78 | ||
| BC094381.1 | - | 3 - 4 | 450 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alkbh1
Genomic Map