Assay Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC027086.1 | - | 2 - 3 | 105 | 79 | |
| BC050856.1 | - | 5 - 6 | 683 | 79 | ||
| BC094381.1 | - | 4 - 5 | 547 | 79 | ||
More Information
Set Membership: |
|

Alkbh1
Genomic Map