Assay Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102565.1 | NP_001096035.1 | 5 - 6 | 761 | 67 | |
| GenBank mRNA | AK143105.1 | - | 7 - 8 | 971 | 67 | |
| AK144828.1 | - | 5 - 6 | 763 | 67 | ||
| BC027086.1 | - | 3 - 4 | 253 | 67 | ||
| BC050856.1 | - | 6 - 7 | 831 | 67 | ||
| BC094381.1 | - | 5 - 6 | 695 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
neuron migration
placenta development
DNA repair
DNA dealkylation involved in DNA repair
regulation of gene expression
cell differentiation
neuron projection development
RNA repair
negative regulation of neuron apoptotic process
developmental growth
positive chemotaxis
oxidative demethylation
DNA demethylation
nucleic acid phosphodiester bond hydrolysis
DNA-(apurinic or apyrimidinic site) lyase activity
protein binding
ferrous iron binding
chemoattractant activity
methylcytosine dioxygenase activity
in utero embryonic development
neuron migration
placenta development
DNA repair
DNA dealkylation involved in DNA repair
regulation of gene expression
cell differentiation
neuron projection development
RNA repair
negative regulation of neuron apoptotic process
developmental growth
positive chemotaxis
oxidative demethylation
DNA demethylation
nucleic acid phosphodiester bond hydrolysis
DNA-(apurinic or apyrimidinic site) lyase activity
protein binding
ferrous iron binding
chemoattractant activity
methylcytosine dioxygenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103515 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146350 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118917 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC027086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC050856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | BC094381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | AK146582.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P522023 | NM_001102565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P126596 | AK042102.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28616730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09761266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09915019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10087712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09923589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09940674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09940676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10389839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10017416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10271318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09940672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09892780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09884525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10369388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09884513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28616695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09929790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10401801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401799 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438681_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434660_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10401795 | - | - |
| Mouse Genome 430 2.0 Array | 1434660_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434660_at | - | - |
| Mouse Genome 430 2.0 Array | 1438681_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438681_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200014777.mm.1 | NM_001102565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014777.mm.1 | NM_001102565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014777.mm.1 | TC1200002125.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200014780.mm.1 | NM_001102565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014780.mm.1 | NM_001102565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014780.mm.1 | TC1200002125.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200014776.mm.1 | NM_001102565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014776.mm.1 | NM_001102565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014776.mm.1 | TC1200002125.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200014778.mm.1 | NM_001102565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014778.mm.1 | NM_001102565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200014778.mm.1 | TC1200002125.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434660_PM_at | - | - |
| Mouse Expression Set 430 | 1438681_at | - | - |
| Mouse Expression Set 430 | 1438681_PM_at | - | - |
| Mouse Expression Set 430 | 1434660_at | - | - |
| Murine Genome U74v2 Set | 167408_PM_at | - | - |
| Murine Genome U74v2 Set | 167408_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438681_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438681_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434660_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434660_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 564223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101580180 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2424932 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002321.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101580180 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101580180 | - | - |

Alkbh1
Genomic Map