Assay Details
| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142580.1 | NP_001136052.1 | 16 - 17 | 1313 | 61 | |
| NM_001142581.1 | NP_001136053.1 | 15 - 16 | 1256 | 61 | ||
| NM_134044.3 | NP_598805.2 | 15 - 16 | 1270 | 61 | ||
| GenBank mRNA | AK046322.1 | - | 16 - 17 | 1284 | 61 | |
| AK049643.1 | - | 16 - 17 | 1260 | 61 | ||
| AK079055.1 | - | 16 - 17 | 1260 | 61 | ||
| AK082261.1 | - | 16 - 17 | 1241 | 61 | ||
| BC023716.1 | - | 15 - 16 | 1281 | 5346489 | 61 | |
More Information
Set Membership: |
|

Vipas39
Genomic Map