Assay Details
| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142580.1 | NP_001136052.1 | 5 - 6 | 510 | 88 | |
| NM_001142581.1 | NP_001136053.1 | 5 - 6 | 510 | 88 | ||
| NM_134044.3 | NP_598805.2 | 5 - 6 | 524 | 88 | ||
| GenBank mRNA | AK038165.1 | - | 5 - 6 | 478 | 88 | |
| AK049643.1 | - | 5 - 6 | 457 | 88 | ||
| AK079055.1 | - | 5 - 6 | 457 | 88 | ||
| AK082261.1 | - | 5 - 6 | 438 | 88 | ||
| AK145043.1 | - | 493 | 88 | |||
| BB610654.1 | - | 5 - 6 | 511 | 88 | ||
| BC016646.1 | - | 5 - 6 | 433 | 2651233 | 88 | |
| BC023716.1 | - | 5 - 6 | 535 | 5346489 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vipas39
Genomic Map