Assay Details
| Gene Symbol: Ercc6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013608.2 | NP_001013626.2 | 11 - 12 | 1905 | 149 | |
| NM_023507.3 | NP_075996.2 | 11 - 12 | 1891 | 149 | ||
| GenBank mRNA | AF217319.1 | - | 10 - 11 | 1708 | 149 | |
| AK049874.1 | - | 11 - 12 | 2217 | 149 | ||
| BC032964.1 | - | 7 - 8 | 919 | 149 | ||
| CN457854.1 | - | 5 - 6 | 553 | 149 | ||
| CO430774.1 | - | 5 - 6 | 538 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ercc6l2
Genomic Map