Assay Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098222.1 | NP_001091692.1 | 3 - 4 | 378 | 56 | |
| NM_198161.2 | NP_937804.1 | 3 - 4 | 388 | 56 | ||
| GenBank mRNA | AK143861.1 | - | 3 - 4 | 311 | 56 | |
| AK157023.1 | - | 3 - 4 | 342 | 56 | ||
| AK183392.1 | - | 1 - 2 | 85 | 56 | ||
| AK214741.1 | - | 1 - 2 | 44 | 56 | ||
| BB863588.1 | - | 3 - 4 | 376 | 56 | ||
| CF729268.1 | - | 3 - 4 | 318 | 56 | ||
| CK625652.1 | - | 3 - 4 | 377 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143861.1, AK157023.1, NM_001098222.1, NM_198161.2 transcripts.
Set Membership: |
|

Bhlhb9
Genomic Map