| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00008259.mm.1 |
NM_001098222.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008259.mm.1 |
NM_001098222.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008259.mm.1 |
NM_198161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008259.mm.1 |
NM_198161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008259.mm.1 |
TC0X00003402.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00008257.mm.1 |
NM_001098222.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008257.mm.1 |
NM_001098222.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008257.mm.1 |
NM_198161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008257.mm.1 |
NM_198161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008257.mm.1 |
TC0X00003402.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00008258.mm.1 |
NM_001098222.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008258.mm.1 |
NM_001098222.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008258.mm.1 |
NM_198161.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008258.mm.1 |
NM_198161.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00008258.mm.1 |
TC0X00003402.mm.1 |
MTA TC |