Assay Details
| Gene Symbol: Rdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021557.5 | NP_067532.2 | 2 - 3 | 213 | 65 | |
| GenBank mRNA | AB030503.1 | - | 2 - 3 | 545 | 65 | |
| AB030504.1 | - | 2 - 3 | 293 | 65 | ||
| AB035959.1 | - | 2 - 3 | 221 | 65 | ||
| AF474027.1 | - | 2 - 3 | 259 | 65 | ||
| AK004413.1 | - | 2 - 3 | 214 | 65 | ||
| AK135443.1 | - | 2 - 3 | 243 | 65 | ||
| AY039032.1 | - | 2 - 3 | 687 | 65 | ||
| BC018261.1 | - | 2 - 3 | 238 | 4460226 | 65 | |
| BC036273.1 | - | 2 - 3 | 238 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rdh11
Genomic Map