Assay Details
| Gene Symbol: Ccl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021443.3 | NP_067418.1 | 1 - 2 | 129 | 61 | |
| GenBank mRNA | AB023418.1 | - | 1 - 2 | 137 | 61 | |
| AK007942.1 | - | 1 - 2 | 130 | 61 | ||
| BC117101.1 | - | 1 - 2 | 127 | 40126420 | 61 | |
| BC117103.1 | - | 1 - 2 | 127 | 40126422 | 61 | |
| BY707942.1 | - | 1 - 2 | 130 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl8
Genomic Map