Assay Details
| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007957.2 | NP_031983.2 | 4 - 5 | 987 | 59 | |
| GenBank mRNA | AF004211.1 | - | 3 - 4 | 1075 | 59 | |
| AF017734.1 | - | 3 - 4 | 704 | 59 | ||
| AF017735.1 | - | 4 - 5 | 787 | 59 | ||
| AF085715.1 | - | 4 - 5 | 988 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684274 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971094 | AF004211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971094 | AF017734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971094 | AF017735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971094 | AF085715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971094 | NM_007957.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104629 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175528 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971094 | AF004211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971094 | AF017734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971094 | AF017735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971094 | AF085715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971094 | NM_007957.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450248 | AF004211.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450248 | AF017734.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450248 | AF017735.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450248 | AF085715.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450248 | NM_007957.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32684277 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175528 | - | - |
| Whole Mouse Genome G4122A | A_51_P450248 | AF004211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450248 | AF017734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450248 | AF017735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450248 | AF085715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450248 | NM_007957.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125704 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14524801 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15221896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14763793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14763797 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14524800 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15221895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32684276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32684278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14911196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14819703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15221896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14887277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14971597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14655645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14971599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14524801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14731913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14911191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32684295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10606915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606916 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420602_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606914 | - | - |
| Mouse Genome 430 2.0 Array | 1420602_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420602_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13984_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13984_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00019110.mm.1 | NM_007957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019110.mm.1 | NM_007957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019110.mm.1 | TC0X00003046.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420602_a_at | - | - |
| Mouse Expression Set 430 | 1420602_PM_a_at | - | - |
| Murine Genome U74v2 Set | 99906_PM_at | - | - |
| Murine Genome U74v2 Set | 99906_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420602_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420602_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420602_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420602_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 924691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480739 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625674 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2625674 | - | - |
| mouse-6 v1.1 (Illumina) | scl53805.5.1_248-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53805.5.1_248-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2625674 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625674 | - | - |

Esx1
Genomic Map