Assay Details
| Gene Symbol: Dhx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172594.2 | NP_766182.2 | 12 - 13 | 2101 | 68 | |
| GenBank mRNA | BC050229.1 | - | 12 - 13 | 2130 | 68 | |
| BC057112.1 | - | 12 - 13 | 2174 | 6417650 | 68 | |
| BC082319.1 | - | 12 - 13 | 2164 | 6417162 | 68 | |
More Information
Set Membership: |
|

Dhx29
Genomic Map