Assay Details
| Gene Symbol: Dhx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172594.2 | NP_766182.2 | 22 - 23 | 3424 | 60 | |
| GenBank mRNA | AK028342.1 | - | 10 - 11 | 1194 | 60 | |
| AK164583.1 | - | 1 - 2 | 149 | 60 | ||
| BC057112.1 | - | 22 - 23 | 3497 | 6417650 | 60 | |
| BC082319.1 | - | 22 - 23 | 3487 | 6417162 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx29
Genomic Map