Assay Details
| Gene Symbol: Dhx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172594.2 | NP_766182.2 | 23 - 24 | 3520 | 107 | |
| GenBank mRNA | AK028342.1 | - | 11 - 12 | 1290 | 107 | |
| BC057112.1 | - | 23 - 24 | 3593 | 6417650 | 107 | |
| BC082319.1 | - | 23 - 24 | 3583 | 6417162 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx29
Genomic Map