Assay Details
| Gene Symbol: Dhx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172594.2 | NP_766182.2 | 8 - 9 | 1057 | 114 | |
| GenBank mRNA | AK049530.1 | - | 8 - 9 | 1130 | 114 | |
| BC020331.1 | - | 8 - 9 | 1150 | 114 | ||
| BC050229.1 | - | 8 - 9 | 1086 | 114 | ||
| BC057112.1 | - | 8 - 9 | 1130 | 6417650 | 114 | |
| BC082319.1 | - | 8 - 9 | 1120 | 6417162 | 114 | |
More Information
Set Membership: |
|

Dhx29
Genomic Map