Assay Details
| Gene Symbol: Ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081062.1 | NP_001074531.1 | 2 - 3 | 664 | 72 | |
| GenBank mRNA | AK053890.1 | - | 1 - 2 | 1269 | 72 | |
| AK086507.1 | - | 2 - 3 | 639 | 72 | ||
| BC147759.1 | - | 2 - 3 | 640 | 9087484 | 72 | |
| BC147766.1 | - | 2 - 3 | 640 | 9087491 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320122 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164146 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164146 | - | - |
| Whole Mouse Genome G4122A | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291501 | NM_001081062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291501 | AK053890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291501 | AK086507.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29337995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29337996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29337994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29338051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10845560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10843159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10954041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10744776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10845563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29338014 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10407252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407253 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455114_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407251 | - | - |
| Mouse Genome 430 2.0 Array | 1455114_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455114_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218630_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 218630_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300008116.mm.1 | NM_001081062.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008116.mm.1 | NM_001081062.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008116.mm.1 | TC1300001274.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300008117.mm.1 | NM_001081062.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008117.mm.1 | NM_001081062.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008117.mm.1 | TC1300001274.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300008115.mm.1 | NM_001081062.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008115.mm.1 | NM_001081062.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008115.mm.1 | TC1300001274.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455114_PM_at | - | - |
| Mouse Expression Set 430 | 1455114_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455114_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455114_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 497441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736471 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736471 | - | - |

Ccno
Genomic Map