Assay Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010560.3 | NP_034690.3 | 3 - 4 | 349 | 99 | |
| GenBank mRNA | AK170020.1 | - | 4 - 5 | 511 | 99 | |
| BC020454.1 | - | 3 - 4 | 303 | 99 | ||
| BC058679.1 | - | 3 - 4 | 310 | 6834623 | 99 | |
| CJ177012.1 | - | 3 - 4 | 351 | 99 | ||
| M83336.1 | - | 2 - 3 | 154 | 99 | ||
| X62646.1 | - | 2 - 3 | 154 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il6st
Genomic Map