Assay Details
| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145965.2 | NP_666077.2 | 6 - 7 | 579 | 109 | |
| NM_172952.3 | NP_766540.2 | 6 - 7 | 579 | 109 | ||
| GenBank mRNA | AK082353.1 | - | 6 - 7 | 501 | 109 | |
| AK144569.1 | - | 6 - 7 | 582 | 109 | ||
| BC127058.1 | - | 6 - 7 | 501 | 40109771 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gphn
Genomic Map