Assay Details
| Gene Symbol: Ano6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253813.1 | NP_001240742.1 | 7 - 8 | 955 | 69 | |
| NM_175344.4 | NP_780553.2 | 6 - 7 | 892 | 69 | ||
| GenBank mRNA | AK159411.1 | - | 7 - 8 | 906 | 69 | |
| BC060732.1 | - | 6 - 7 | 892 | 6409332 | 69 | |
More Information
Set Membership: |
|

Ano6
Genomic Map