Assay Details
| Gene Symbol: Scaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028148.2 | NP_082424.2 | 3 - 4 | 484 | 110 | |
| GenBank mRNA | AK129478.1 | - | 3 - 4 | 376 | 110 | |
| AK163564.1 | - | 3 - 4 | 346 | 110 | ||
| BC058682.1 | - | 3 - 4 | 465 | 110 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scaf11
Genomic Map