Assay Details
| Gene Symbol: Scaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028148.2 | NP_082424.2 | 9 - 10 | 991 | 94 | |
| GenBank mRNA | AK129478.1 | - | 9 - 10 | 883 | 94 | |
| BC058682.1 | - | 9 - 10 | 972 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scaf11
Genomic Map