Assay Details
| Gene Symbol: D030018L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137238.1 | - | 3 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | anoctamin 6 | Ano6 |
2900059G15Rik, AA407480, AW554778, F730003B03Rik, Tmem16f |
-99.2kb | Anti-sense | Chr. 15 - 95790843 - 95975472 on NCBI Build GRCm38 |
| Downstream |
eukaryotic translation initiation factor 3, subunit I pseudogene |
Gm4371 |
EG667874, Gm8854 |
106.5kb | Anti-sense | Chr. 15 - 96182800 - 96203470 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

D030018L15Rik
Genomic Map