Assay Details
| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081402.1 | NP_001074871.1 | 10 - 11 | 1161 | 71 | |
| GenBank mRNA | AK159714.1 | - | 10 - 11 | 1143 | 71 | |
| AK160933.1 | - | 10 - 11 | 1097 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr70
Genomic Map